3-(2-Naphthyl)-L-alanine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (S)-alpha-Amino-2-naphthalenepropionic Acid, (S)-2-Amino-3-(naphthalen-2-yl)propanoic acid, H-3-(2-Naphthyl)-Ala-OH

H63354.03 H63354.06 H63354.14
H63354.03EA 34.7 GBP
H63354.03 H63354.06 H63354.14
3-(2-Naphthyl)-L-alanine 97%
3-(2-Naphthyl)-L-alanine

Purity: 97% CAS No.: 58438-03-2 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 2726272 Optical Rotation: -11°(c= 1M HCl) MDL No.: MFCD00066087

Formula: C₁₃H₁₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD00066087
CAS Number: 58438-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR