Ethyl-2-methylquinoline-6-carboxylate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H63213.03 H63213.06
H63213.03EA 26.4 GBP
H63213.03 H63213.06
Ethyl-2-methylquinoline-6-carboxylate 97%
Ethyl-2-methylquinoline-6-carboxylate

Purity: 97% CAS No.: 855763-77-8 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Physical Form: Solid MDL No.: MFCD09879697

Formula: C₁₃H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09879697
CAS Number: 855763-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR