Boc-D-Glu-OtBu 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-Boc-D-glutamic acid 1-tert-butyl ester

H62320.03 H62320.06
H62320.03EA 123 GBP
H62320.03 H62320.06
Boc-D-Glu-OtBu 95%
Boc-D-Glu-OtBu

Purity: 95% CAS No.: 73872-71-6 Molecular Formula: C14H25NO6 Formula Weight: 303.35 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale brown MDL No.: MFCD00076926

Formula: C₁₄H₂₅NO₆
MW: 303.355 g/mol
MDL Number: MFCD00076926
CAS Number: 73872-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR