Methyl-4-piperidinecarboxylate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H61754.14 H61754.22
H61754.14EA 43.2 GBP
H61754.14 H61754.22
Methyl-4-piperidinecarboxylate ≥98%
Methyl 4-piperidinecarboxylate
Formula: C₇H₁₃NO₂
Boiling Pt: 85…90 °C
Density: 1.06
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190578
CAS Number: 2971-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR