Phenylacetamide 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H60626.MDEA 35.4 GBP
H60626.MD
Phenylacetamide 99%
Phenylacetamide

Used as a pharmaceutical intermediate.


Store at 4°C.

Formula: C₈H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00059193
CAS Number: 103-81-1
EINECS: 203-147-0

Order Now

Specification Test Results

Assay (HPLC) 98,5%
Solubility Soluble in methanol
Storage temperature 4 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR