5-Bromo-o-toluic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H60516.06
H60516.06EA 90.2 GBP
H60516.06
5-Bromo-o-toluic acid ≥97%
5-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00267350
CAS Number: 79669-49-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR