Methyl-2-methyl-3-nitrobenzoate ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H60242.06 H60242.14
H60242.06EA 31.2 GBP
H60242.06 H60242.14
Methyl-2-methyl-3-nitrobenzoate ≥97%
Methyl 2-methyl-3-nitrobenzoate
Formula: C₉H₉NO₄
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00082564
CAS Number: 59382-59-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR