1-Benzyl-5-bromoindole 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Benzyl-5-bromo-1H-indole

H57676.03 H57676.06
H57676.03EA 50.2 GBP
H57676.03 H57676.06
1-Benzyl-5-bromoindole 97%
1-Benzyl-5-bromoindole

Purity: 97% CAS No.: 10075-51-1 Molecular Formula: C15H12BrN Formula Weight: 286.17 MDL No.: MFCD04337704

Formula: C₁₅H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD04337704
CAS Number: 10075-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR