(R)-(-)-Nipecotic acid ≥96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H57434.03 H57434.06 H57434.14
H57434.03EA 39.8 GBP
H57434.03 H57434.06 H57434.14
(R)-(-)-Nipecotic acid ≥96%
(R)-(-)-Nipecotic acid

Purity: 96+% CAS No.: 25137-00-2 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: 250-255° Optical Rotation: -5° (c=1 in water)

Formula: C₆H₁₁NO₂
Melting Pt: 250…255 °C
Storage Temperature: Ambient
MDL Number: MFCD01630787
CAS Number: 25137-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR