2,4-Dichloro-5-methylpyrimidine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H55206.03 H55206.06
H55206.03EA 43.2 GBP
H55206.03 H55206.06
2,4-Dichloro-5-methylpyrimidine ≥98%
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
Boiling Pt: 108…109 °C (11 mmHg)
Melting Pt: 26…28 °C
Density: 1.39
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR