Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H55020.03 H55020.06
H55020.03EA 0 GBP
H55020.03 H55020.06
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) ≥97%
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II)

Purity: 97% CAS No.: 18498-01-6 Molecular Formula: C26H24Cl2CoP2 Formula Weight: 528.26 Melting Point: 260° dec. MDL No.: MFCD00134173

Formula: C₂₆H₂₄Cl₂CoP₂
Melting Pt: 260 °C (dec.)
MDL Number: MFCD00134173
CAS Number: 18498-01-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR