Methyl-2-amino-5-methoxybenzoate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl 2-amino-5-methoxybenzoate

H54704.MD H54704.03 H54704.06
H54704.MDEA 42.6 GBP
H54704.MD H54704.03 H54704.06
Methyl-2-amino-5-methoxybenzoate 96%
Methyl 2-amino-5-methoxybenzoate

Purity: 96% CAS No.: 2475-80-1 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 32-33° MDL No.: MFCD09038482

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 32…33 °C
Storage Temperature: Ambient
MDL Number: MFCD09038482
CAS Number: 2475-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR