6-Amino-5-methoxypyridine-3-boronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

H54546.03
H54546.03EA 210 GBP
H54546.03
6-Amino-5-methoxypyridine-3-boronic acid pinacol ester 96%
6-Amino-5-methoxypyridine-3-boronic acid pinacol ester

Purity: 96% CAS No.: 947249-19-6 Molecular Formula: C12H19BN2O3 Formula Weight: 250.10 Melting Point: 98-103° MDL No.: MFCD11977248

Formula: C₁₂H₁₉BN₂O₃
MW: 250.10 g/mol
Melting Pt: 98…103 °C
Storage Temperature: Ambient
MDL Number: MFCD11977248
CAS Number: 947249-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR