Boc-(S)-thiazolidine-2-carboxylic acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H52793.03 H52793.06
H52793.03EA 74.2 GBP
H52793.03 H52793.06
Boc-(S)-thiazolidine-2-carboxylic acid 97%
Boc-(S)-thiazolidine-2-carboxylic acid

Purity: 97% CAS No.: 891192-95-3 Molecular Formula: C9H15NO4S Formula Weight: 233.28 Melting Point: 90-93° Optical Rotation: -47° (c=5 in methanol) MDL No.: MFCD02682342

Formula: C₉H₁₅NO₄S
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD02682342
CAS Number: 891192-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR