Boc-D-Tyr(ME)-OH 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

H52082.03 H52082.06
H52082.03EA 42.4 GBP
H52082.03 H52082.06
Boc-D-Tyr(ME)-OH 95%
Boc-D-Tyr(ME)-OH

Purity: 95% CAS No.: 68856-96-2 Molecular Formula: C15H21NO5 Formula Weight: 295.33

Formula: C₁₅H₂₁NO₅
MW: 295.335 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065604
CAS Number: 68856-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR