DSS (Disuccinimidyl suberate) ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) octanedioate, Suberic acid bis(N-hydroxysuccinimide ester), Disuccinimidyl octanedioate, DSS

H51771.ME H51771.03
H51771.MEEA 44.2 GBP
H51771.ME H51771.03
DSS (Disuccinimidyl suberate) ≥97%
DSS (Disuccinimidyl suberate)

Purity: 97% CAS No.: 68528-80-3 Molecular Formula: C16H20N2O8 Formula Weight: 368.34 Melting Point: 166-170° MDL No.: MFCD00049059

Formula: C₁₆H₂₀N₂O₈
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00049059
CAS Number: 68528-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR