Platinum 5% on activated carbon paste Type 128M

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Platinium

H37641.06 H37641.14
H37641.06EA 69.8 GBP
H37641.06 H37641.14
Platinum 5% on activated carbon paste Type 128M
Platinum

CAS No.: 7440-06-4 Physical Form: Solid Appearance: Black Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR