4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H34429.14 H34429.03 H34429.06
H34429.14EA 141 GBP
H34429.14 H34429.03 H34429.06
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione

Purity: 97% CAS No.: 582-65-0 Molecular Formula: C10H6F4O2 Formula Weight: 234.15 Melting Point: 40-44° MDL No.: MFCD03419766

Formula: C₁₀H₆F₄O₂
Melting Pt: 40…44 °C
Storage Temperature: Ambient
MDL Number: MFCD03419766
CAS Number: 582-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR