3-Chloro-5-(trifluoromethyl)picolinic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid

H34258.03
H34258.03EA 86.7 GBP
H34258.03
3-Chloro-5-(trifluoromethyl)picolinic acid ≥97%
3-Chloro-5-(trifluoromethyl)picolinic acid
Formula: C₇H₃ClF₃NO₂
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00277482
CAS Number: 80194-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR