Diethyl ether stabilised for spectrophotometry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

H33678.AP H33678.AU H33678.0F
H33678.APEA 52.4 GBP
H33678.AP H33678.AU H33678.0F
Diethyl ether stabilised for spectrophotometry
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR