Methyl-2-chloro-6-methoxyisonicotinate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate

H33438.06 H33438.14
H33438.06EA 65.7 GBP
H33438.06 H33438.14
Methyl-2-chloro-6-methoxyisonicotinate ≥98%
Methyl 2-chloro-6-methoxyisonicotinate

Purity: 98% CAS No.: 42521-10-8 Molecular Formula: C8H8ClNO3 Formula Weight: 201.61 Melting Point: 103-105° MDL No.: MFCD00173929

Formula: C₈H₈ClNO₃
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00173929
CAS Number: 42521-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR