Ethyl-1H-indazole-3-carboxylate 95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ethyl 1H-indazole-3-carboxylate

H32098.03
H32098.03EA 247 GBP
H32098.03
Ethyl-1H-indazole-3-carboxylate 95%
Ethyl 1H-indazole-3-carboxylate

Purity: 95% CAS No.: 4498-68-4 Molecular Formula: C10H10N2O2 Formula Weight: 190.20 Melting Point: 136-138° MDL No.: MFCD01138134

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD01138134
CAS Number: 4498-68-4
EINECS: 224-795-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR