2,3-Dimethyl-4-fluoroaniline ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H31560.MD H31560.03
H31560.MDEA 55.3 GBP
H31560.MD H31560.03
2,3-Dimethyl-4-fluoroaniline ≥97%
2,3-Dimethyl-4-fluoroaniline

Purity: 97% CAS No.: 1737-68-4 Molecular Formula: C8H10FN Formula Weight: 139.17 Melting Point: 42-46° UN No.: 2811 MDL No.: MFCD04972870

Formula: C₈H₁₀FN
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD04972870
CAS Number: 1737-68-4
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR