2,3-Difluoro-4-hydroxybenzaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H31538.03
H31538.03EA 124 GBP
H31538.03
2,3-Difluoro-4-hydroxybenzaldehyde ≥97%
2,3-Difluoro-4-hydroxybenzaldehyde

Purity: 98% CAS No.: 676500-39-3 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 140-144° MDL No.: MFCD08235194

Formula: C₇H₄F₂O₂
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD08235194
CAS Number: 676500-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR