2-Amino-5-bromothiazole hydrobromide 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr

H31514.03 H31514.06 H31514.14
H31514.03EA 18.9 GBP
H31514.03 H31514.06 H31514.14
2-Amino-5-bromothiazole hydrobromide 97%
2-Amino-5-bromothiazole hydrobromide

Purity: 97% CAS No.: 61296-22-8 Molecular Formula: C3H3BrN2S.HBr Formula Weight: 259.95 Melting Point: ca 165° dec. MDL No.: MFCD00012712

Formula: C₃H₃BrN₂S
MW: 259.95 g/mol
Melting Pt: ∼ 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012712
CAS Number: 61296-22-8
EINECS: 262-699-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR