2,2'-Bis(diphenylphosphino)biphenyl 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: BIPHEP, 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl

H31306.MD H31306.03 H31306.06
H31306.MDEA 54.2 GBP
H31306.MD H31306.03 H31306.06
2,2'-Bis(diphenylphosphino)biphenyl 98%
2,2'-Bis(diphenylphosphino)biphenyl

Purity: 98% CAS No.: 84783-64-2 Molecular Formula: C36H28P2 Formula Weight: 522.57 Melting Point: 211-215° MDL No.: MFCD03094574

Formula: C₃₆H₂₈P₂
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD03094574
CAS Number: 84783-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR