tert-Butyl vinyl ether 98% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with N,N-diethylaniline ∼ 0.1%

H30770.06 H30770.14
H30770.06EA 46.3 GBP
H30770.06 H30770.14
tert-Butyl vinyl ether 98% stabilised
tert-Butyl vinyl ether

Stab. with ca 0.1% N,N-diethylaniline


Purity: 98% CAS No.: 926-02-3 Molecular Formula: C6H12O Formula Weight: 100.16 Storage Temperature: 2°C to 8°C Flash Point: -17°(1°F) Refractive Index: 1.3990 Odour: Characteristic Boiling Point: 75-76° UN No.: UN3271 MDL No.: MFCD00048246

Formula: C₆H₁₂O
Boiling Pt: 75…76 °C
Density: 0.762
Flash Pt: –17 °C (1 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00048246
CAS Number: 926-02-3
EINECS: 618-860-6
UN: 3271
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR