Hydrogen chloride 1 M in diethyl ether

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H27125.AE H27125.AP
H27125.AEEA 56.5 GBP
H27125.AE H27125.AP
Hydrogen chloride 1 M in diethyl ether
Hydrogen chloride

CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Storage Temperature: 2°C to 8°C Flash Point: -40°(-40°F) Refractive Index: 1.3535 UN No.: 2924 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46 g/mol
Density: 0.731
Flash Pt: –40 °C (–40 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR