3,6-Dimethoxypyridazine-4-carboxylic acid 96%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H26877.03
H26877.03EA 70.2 GBP
H26877.03
3,6-Dimethoxypyridazine-4-carboxylic acid 96%
3,6-Dimethoxypyridazine-4-carboxylic acid

Purity: 96% CAS No.: 89694-24-6 Molecular Formula: C7H8N2O4 Formula Weight: 184.15 MDL No.: MFCD03426073

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426073
CAS Number: 89694-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR