3-(3,4-Difluorophenyl)propionic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H26185.03 H26185.06
H26185.03EA 14.6 GBP
H26185.03 H26185.06
3-(3,4-Difluorophenyl)propionic acid ≥98%
3-(3,4-Difluorophenyl)propionic acid

Purity: 98% CAS No.: 161712-75-0 Molecular Formula: C9H8F2O2 Formula Weight: 186.16 Melting Point: 49-53° MDL No.: MFCD00799520

Formula: C₉H₈F₂O₂
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00799520
CAS Number: 161712-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR