HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H26082.03 H26082.06 H26082.14
H26082.03EA 27.6 GBP
H26082.03 H26082.06 H26082.14
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

Purity: 99% CAS No.: 148893-10-1 Molecular Formula: C10H15F6N6OP Formula Weight: 380.23 Melting Point: ca 185° dec. MDL No.: MFCD00274639

Formula: C₁₀H₁₅F₆N₆OP
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00274639
CAS Number: 148893-10-1
UN: 1325
ADR: 4.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR