(4-Chlorophenyl)methanesulphonyl chloride ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Chloro-α-toluenesulphonyl chloride

H26017.03 H26017.06
H26017.03EA 50.4 GBP
H26017.03 H26017.06
(4-Chlorophenyl)methanesulphonyl chloride ≥97%
(4-Chlorophenyl)methanesulphonyl chloride

Purity: 97% CAS No.: 6966-45-6 Molecular Formula: C7H6Cl2O2S Formula Weight: 225.10 Melting Point: 93-96° UN No.: 3261 MDL No.: MFCD01631927

Formula: C₇H₆Cl₂O₂S
MW: 225.10 g/mol
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD01631927
CAS Number: 6966-45-6
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR