Lead acetate cotton, StandARd®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H220-03EA 484 GBP
H220-03
Lead acetate cotton, StandARd®, Macron Fine Chemicals™
Lead acetate cotton

In cotton balls. For arsenic.

Formula: (H₃CCOO)₂Pb
Melting Pt: 175…180 °C
Density: ∼2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 6080-56-4
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR