Coronavirus and SARS-CoV-2 ELISA kits, NovaLisa®
Supplier: GOLD STANDARD DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
COVA0940 COVG0940 COVM0940
GSDCCOVA0940EA 170 GBP
GSDCCOVA0940 GSDCCOVG0940 GSDCCOVM0940
Coronavirus and SARS-CoV-2 ELISA kits, NovaLisa®
Clinical Diagnostic Supplies Clinical Reagents and Kits
This ELISA kit is intended for the qualitative detection of IgA class antibodies to SARS-CoV-2 virus in human serum and plasma for diagnostics of exposure to coronavirus (COVID-19).
  • Separate detection of IgG, IgM and IgA antibodies
  • Advantages of the NovaTec reagent system
  • Total incubation under 2 hours
Compatible with diverse open platform ELISA analysers.
Certifications: CE-IVD marked.