Paediatric tubes with cap, Vacuette®
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
459036_100 459075_100 459084_100 459085_100
GREI459036_100EA 46.2 GBP
GREI459036_100 GREI459075_100 GREI459084_100 GREI459085_100
Paediatric tubes with cap, Vacuette®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clar.
The innovative VACUETTE® blood collection tubes made from practically unbreakable plastic have become well established on the market. They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.