Blood collection tubes, with EDTA, VACUETTE®
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
454034_1200 454020_1200 454021_1200 454023_1200 454036_1200 454047_1200 454086_1200 454087_1200 455036_1200 456036_1200 456023_1200 455040_1200 454222_1200 456038_1200 454217_1200 454024_1200 455045_1200 454209_1200
391-3760EA 165 GBP
391-3760 GREI454020_1200 GREI454021_1200 GREI454023_1200 GREI454036_1200 GREI454047_1200 GREI454086_1200 GREI454087_1200 GREI455036_1200 GREI456036_1200 GREI456023_1200 GREI455040_1200 GREI454222_1200 GREI456038_1200 GREI454217_1200 GREI454024_1200 GREI455045_1200 GREI454209_1200
Blood collection tubes, with EDTA, VACUETTE®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
EDTA tubes are ideal for the examination of whole blood in haematology and are offered as either K2EDTA or K3EDTA tubes.
  • EDTA binds the calcium ions and therefore blocks the coagulation cascade
Order Now
SPECIFICATIONS
Environmentally Preferable