Serum collection tubes, VACUETTE®
Supplier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
454028_1200 454067_1200 454071_1200 454092_1200 454096_1200 454214_1200 455071_1200 455092_1200 456010_1200 456018_1200 456071_1200 456073_1200 456089_1200 456092_1200 454098_1200 454204_1200 454236_1200 454228_1200
GREI454028_1200EA 226 GBP
GREI454028_1200 GREI454067_1200 GREI454071_1200 GREI454092_1200 GREI454096_1200 GREI454214_1200 GREI455071_1200 GREI455092_1200 GREI456010_1200 GREI456018_1200 GREI456071_1200 GREI456073_1200 GREI456089_1200 GREI456092_1200 GREI454098_1200 GREI454204_1200 GREI454236_1200 GREI454228_1200
Serum collection tubes, VACUETTE®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
VACUETTE® serum tubes are coated with micronised silica particles which activate clotting when tubes are gently inverted. These tubes are used for routine clinical chemistry, immunohematology, serology and immunology testing.
These serum tubes with gel contain a barrier gel that is present in the bottom of the tube. During centrifugation, the barrier gel moves upward to the serum-clot interface where it forms a stable barrier, separating the serum from the cells.
Certifications: FDA 510 (k) cleared for therapeutic drug monitoring (TDM).
Caution: *The product(s) on this page areMore Product Information