Trays and Inserts
Supplier: GRATNELL'S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LF1 F0120 F0106 F0110 F0206 F0210 F0206P6 F0105P12 F2501P6 F2509P6 F2510P6 IM03F0119P6 IM08F0119P6 F0220 F0106P12 F0109 F0209 F0209P6
GRATLF1EA 710 GBP
GRATLF1 GRATF0120 GRATF0106 GRATF0110 GRATF0206 GRATF0210 GRATF0206P6 GRATF0105P12 GRATF2501P6 GRATF2509P6 GRATF2510P6 GRATIM03F0119P6 GRATIM08F0119P6 GRATF0220 GRATF0106P12 GRATF0109 GRATF0209 GRATF0209P6
Trays and Inserts
Trays/Dishes/Pots Drawer Organisation Trays
These all adds up to a comprehensive storage system.
  • Available in a range of shapes and sizes
  • Available in many colours
  • Rigid and foam inserts available
  • Lid available to cover contents
These award-winning trays come in a range of shapes and sizes to fit most standard school furniture. Made from tough, recyclable polypropylene, these trays will last for many years. Standard F trays are available in a wide variety of colors to suit most color schemes. These F trays are designed to fit 450 mm deep furniture. Available in shallow, deep, extra deep andMore Product Information