Refrigerated/heating circulating baths, ecocool™ series
Supplier: GRANT INSTRUMENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ECOCOOL150R ECOCOOL100R
GRANECOCOOL150REA 4538 GBP
GRANECOCOOL150R GRANECOCOOL100R
Refrigerated/heating circulating baths, ecocool™ series
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
A range of eco-friendly, refrigerated/heating circulating baths with an intuitive user-interface and sleek design. Units offer significant running cost savings whilst delivering powerful cooling when needed. Choice of 2 models with temperature range of –25 to +150 °C (model dependent*). Ideal for applications in pharmaceutical, education, industrial and life science areas for use with mini pilot plant reactors, cooling rotary evaporators, replacing running tap water cooling, immersing small samples, photometry, chromatography systems, sample preparation, general cooling, reaction chemistry,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable