IIRR21010203441 : BioCompact IIRR210 Underbench Refrigerator, 2 to 20?C, white exterior, solid door, ABS plastic interior, door lock. Left or right door hinges. Skids front/recessed wheels rear. Programmable controller, interior & exterior ATEX compl 1 * 1 items
Supplier: GRAM
IIRR21010203441 : BioCompact IIRR210 Underbench Refrigerator, 2 to 20?C, white exterior, solid door, ABS plastic interior, door lock. Left or right door hinges. Skids front/recessed wheels rear. Programmable controller, interior & exterior ATEX compl 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.