Bottles, narrow neck, rectangular, PP, with screw caps, Corning® Gosselin™
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FV250-02 FV500-09
GOSSFV250-02EA 528 GBP
GOSSFV250-02 GOSSFV500-09
Bottles, narrow neck, rectangular, PP, with screw caps, Corning® Gosselin™
Bottles Plastic Bottles
Polypropylene, translucent, graduated with red tamper evident HDPE screw caps.
  • Rectangular cross section bottles save storage space
  • Graduated bottles are supplied with caps but are unassembled
  • Bottles are sterile, irradiated and are supplied capped
Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.