Racks for pipette boxes
Supplier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P04 P06
GLWAP04EA 269 GBP
GLWAP04 GLWAP06
Racks for pipette boxes
Pipette Stands
These stainless steel racks are suitable for the storage of four or six GLW pipette boxes (A30, A40, A45 / S30, S40, S45).
Ordering information: Pipette boxes are not included.
Order Now
SPECIFICATIONS
Environmentally Preferable