Coloured slide boxes
Supplier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K25B K25G K25O K25R K25S K25T K25V K25W K25Y
GLWAK25BEA 12.6 GBP
GLWAK25B GLWAK25G GLWAK25O GLWAK25R GLWAK25S GLWAK25T GLWAK25V GLWAK25W GLWAK25Y
Coloured slide boxes
Slide Boxes
Slide boxes with 25 compartments and snap lock provide a secure hold for the slides and coverslips.
  • Stackable for use as a minimum space as possible
  • Can be used in a freezer down to –80 °C
  • Numbers in the bottom of the box and hinged lid
Designed for storing, transporting, and freezing the samples.
Ordering information: Dividers must be ordered separately.