Analytical HPLC columns, Inertsil®
Supplier: GL Sciences B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5020-01901 5020-05124 5020-04942 5020-05546 5020-04612 5020-03946 5020-03936 5020-04014 5020-04012 5020-01762 5020-01821 5020-01822 5020-01812 5020-01811 5020-01802 5020-01801 5020-01771 5020-01941 5020-01912 5020-30146 5020-30125 5020-30144 5020-30145 5020-30142
GLSC5020-01901EA 1260 GBP
GLSC5020-01901 GLSC5020-05124 GLSC5020-04942 GLSC5020-05546 GLSC5020-04612 GLSC5020-03946 GLSC5020-03936 GLSC5020-04014 GLSC5020-04012 GLSC5020-01762 GLSC5020-01821 GLSC5020-01822 GLSC5020-01812 GLSC5020-01811 GLSC5020-01802 GLSC5020-01801 GLSC5020-01771 GLSC5020-01941 GLSC5020-01912 GLSC5020-30146 GLSC5020-30125 GLSC5020-30144 GLSC5020-30145 GLSC5020-30142
Analytical HPLC columns, Inertsil®
Chromatography Columns
Inertsil® series phases are based on a purer, higher surface area silica which is specially manufactured to provide maximum phase coverage. The result is a series of columns which provide excellent peak shapes using simple eluents while operating at low pressure.
Inertsil® ODS-3 is used widely and reliably for long established methods in pharmaceutical, and contract research labs. It has a relatively strong retentivity compared to commercially available ODS columns.
Inertsil® ODS-3V offers all of the outstanding chromatographic benefits of Inertsil® ODS-3 with the added benefit of a moreMore Product Information