Tissue grinders, round
Supplier: GLASS PRECISION ENGINEERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20701J 20706J 20707J 20708J 20709J 20710J 20702J 20703J 20704J 20705J 20711J 20712J 20713J 20714J 20715J 20716J 20717J 20718J 20719J 20720J
GLPR20701JEA 146 GBP
GLPR20701J GLPR20706J GLPR20707J GLPR20708J GLPR20709J GLPR20710J GLPR20702J GLPR20703J GLPR20704J GLPR20705J GLPR20711J GLPR20712J GLPR20713J GLPR20714J GLPR20715J GLPR20716J GLPR20717J GLPR20718J GLPR20719J GLPR20720J
Tissue grinders, round
Homogenisers Tissue Grinders
This all-glass Uni-Form range of round body tissue grinders is made from sterilisable heat-resisting glass. Bodies are available in clear precision glass or ground glass for more effective homogenisation of tougher tissues.
  • Rod ends for standard manual use, or hollow pestles Tenbroeck Style for use with coolants/dry ice
  • Grinding clearance pestle and mortar is 0,152 to 0,254 mm