Tissue grinders, Mini series
Supplier: GLASS PRECISION ENGINEERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20404 20405 20406 20402 20403
GLPR20404EA 82.5 GBP
GLPR20404 GLPR20405 GLPR20406 GLPR20402 GLPR20403
Tissue grinders, Mini series
Homogenisers Tissue Grinders
These mini homogenisers are made of high quality borosilicate glass and are ideal for micro tissue work. The mortar is made from clear precision glass and pestles have ground glass working length. Mini homogenisers are available with or without handles.
Grinding clearance pestle and mortar: 0,152 to 0,254 mm.