Pump heads for peristaltic pumps, MINIPuls® 3
Supplier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F117604 F117606 F117608 F117800 F117830 F117831
GILSF117604EA 666 GBP
GILSF117604 GILSF117606 GILSF117608 GILSF117800 GILSF117830 GILSF117831
Pump heads for peristaltic pumps, MINIPuls® 3
Pumps Pump Heads Peristaltic Pump Heads
Add channels to your MINIPULS® 3 or Evolution and allow even greater flexibility in your laboratory.
Order Now
SPECIFICATIONS
Model R1 R4 R8 R2 R2/HF R4/HF
Environmentally Preferable
No. of channels 1 4 8 2 2 4