Safe aspiration station
Supplier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F110744 F110746
GILSF110744EA 2186 GBP
GILSF110744 GILSF110746
Safe aspiration station
Pumps Vacuum Aspiration Systems
The Gilson® safe aspiration station is a stand-alone motorised vacuum system for quick and easy aspiration of biological liquid waste disposal. Liquids are quickly and safely aspirated and collected in a 4 L polypropylene bottle. The station offers adjustable aspiration speed and handles, ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.
  • Safe and contamination-free
  • Adaptable
  • Convenient
The closed, stand-alone unit is designed to prevent contamination hazards. All parts in contact with liquidsMore Product Information