Haematoxylin solution_SC Gill I, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

MQ500

GHS132-1L
GHS132-1LEA 82.3 GBP
GHS132-1L
Haematoxylin solution_SC Gill I, Sigma-Aldrich®
Haematoxylin staining solutions

Gill no.1 formulation is used as a progressive cytology stain.


  • Used with hematoxylin and eosin staining
  • Contains 2 g/l certified hematoxylin

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca68de6f2bec1bf4a4e88490e589eb99f908fb1f&vid=57a749a32e5531a65a02d02807ed843ececa2a4b&prid=web&clid=SAPDAM IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR