Digestion units for Kjeldahl digestion, KJELDATHERM® KT/KT-L
Supplier: GERHARDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
12-0700 12-0702 12-0704 12-0706 12-0725 12-0727 12-0729 12-0731 12-0743 12-0745
GERH12-0700EA 0 GBP
GERH12-0700 GERH12-0702 GERH12-0704 GERH12-0706 GERH12-0725 GERH12-0727 GERH12-0729 GERH12-0731 GERH12-0743 GERH12-0745
Digestion units for Kjeldahl digestion, KJELDATHERM® KT/KT-L
Digestion Systems Mineralisation Workstations
These high-precision, programmable digestion units are ideal for Kjeldahl digestion in 100, 250 or 400 ml digestion tubes. Depending on the sample volume, you can choose between 8-, 20- or 40- place systems.
  • Aluminium digestion block with optimum energy utilisation and effective all-round insulation
  • Sample tubes and extraction unit made of high quality DURAN® glass
  • Automatic control of TURBOSOG and cooling water valve
  • Illuminated digestion chamber with enamelled, acid-resistant surface
  • Pre-heating function for warming up the digestion block
  • Data
More Product Information